Principaux documents
Z677272
Dermatril nitrile gloves
size M
Synonyme(s) :
Chemical Resistant Nitrile Gloves, Nitrile Laboratory Gloves, disposable glove
About This Item
Code UNSPSC :
46181504
Nomenclature NACRES :
NB.35
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
blue gloves
blue nitrile
Taille
M
Stérilité
non-sterile
Caractéristiques
CE compliant
latex free
powder-free
Conditionnement
pack of 100 ea (boxes of 100)
Fabricant/nom de marque
KCL 740
L × épaisseur
250 mm × 0.11 mm
Description générale
Dermatril is a blue nitrile, powder-free, disposable glove providing a high level of protection against chemical and bacteriological contamination. The Dermatril disposable glove also displays very good physical durability in comparison to conventional disposable gloves.
All Dermatril disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
Caractéristiques et avantages
  • Anti-static.
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea.
  • Roughened fingertips for better grip.
  • Rolled cuff edge for easier donning.
  • CE compliant.
  • Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 455, EN 61340-5-1, ASTM 1671:2001
Informations légales
Dermatril is a trademark of KCL GmbH
Certificats d'analyse (COA)
Lot/Batch Number
Désolés, nous n'avons pas de COA pour ce produit disponible en ligne pour le moment.
Questions
1–2 of 2 Questions  
  1. Will these glove block 1,2 Dimethoxy ethane?
    1 answer
    1. Glove recommendations available in section 8 of the material SDS. For 1,2-dimethoxyethane product 25927, the recommended glove materials are Viton and Nitrile Rubber.
      Helpful?
  2. will these block toluene
    1 answer
    1. Yes, these nitrile gloves meet EN 374 (cat. III), EN 388 (cat. II), EN 455, EN 61340-5-1, and ASTM 1671:2001 standards and are suitable for handling toluene.