Key Documents
Z672947
BRAND® accu-jet® pro silicone replacement adapter
valve L × W × H 2 in. × 0.5 in. × 0.5 in., PTFE valve (non-return)
Synonym(s):
Tubing Connectors
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
PTFE valve (non-return)
manufacturer/tradename
BRAND 26508
packaging
1 ea of
valve L × W × H
2 in. × 0.5 in. × 0.5 in.
Related Categories
General description
The BRAND® accu-jet® pro silicone pipette adapter features a reliable PTFE check valve, ensuring efficient liquid transfer without backflow. Compact in size (L 2" x W 0.5" x H 0.5"), this non-return valve replacement adapter enhances the functionality of your pipette while providing a secure fit.
Features and Benefits
  • Silicone pipette adapter with PTFE check valve
  • Dimensions: L 2" x W 0.5" x H 0.5"
  • Non-return valve
Other Notes
BR26508 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
accu-jet is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)