Skip to Content
Merck
All Photos(1)

Key Documents

Z355445

Millipore

PetriSlides, Millipore®

size 51 mm × 76 mm, polystyrene

Synonym(s):

PetriSlides

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.14
Pricing and availability is not currently available.

material

polystyrene

sterility

non-sterile

manufacturer/tradename

Millipore PDMA04700

size

51 mm × 76 mm

color

clear

General description

PetriSlides provide a storage and examination container for 47 mm diameter test filters. Substitute for Petri Dishes. Holds filter securely in place. Transparent cover allows microscopic examination without removal. Rectangular base has rounded corners for mounting on microscope stage.

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service