BR723015
BRAND® Haemacytometer cover glass, for counting chambers
borosilicate glass, size 20 mm × 26 mm × 0.4 mm
Synonym(s):
Cover glasses, Microscopy
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
feature
IVD
manufacturer/tradename
BRAND 723015
size
20 mm × 26 mm × 0.4 mm
binding type
non-treated surface
General description
Borosilicate glass, DIN ISO 8255. Refractive index ne = 1.52 ± 0.01, Abbe number ve = 56.5 ± 0.5. Flatness tolerance ± 3 μm.
They differ from ordinary cover glasses by their plain ground and polished surface.
They differ from ordinary cover glasses by their plain ground and polished surface.
Packaging
2 cover glasses in a plastic bag, separated by tissue paper.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)