Z290505
Sigma-Aldrich® vacuum filtration assembly
for 47 mm filters with stainless steel screen, #8 stopper, same as Z290432 but without side arm flask
All Photos(2)
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
Recommended Products
material
glass housing
stainless steel membrane
feature
cap plug seal
graduation
packaging
pkg of 1 ea
neck diam.
41 mm
Related Categories
General description
Same as Z290432 but without side arm flask.
Vacuum filtration apparatus for HPLC solvent purification, particulate analysis, bacteriological studies, radiochemical determinations, quantitative gravimetric analysis, and similar applications. Clamp seals filter membrane between flanges on funnel top and funnel stem.
Includes: Funnel top, funnel stem/membrane support, aluminum clamp, stopper (except standard taper units) and filtration flask (where indicated).
For replacement parts, visit our online glassware catalog at www.sigma-aldrich.com
Includes: Funnel top, funnel stem/membrane support, aluminum clamp, stopper (except standard taper units) and filtration flask (where indicated).
For replacement parts, visit our online glassware catalog at www.sigma-aldrich.com
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)