Z290408
Sigma-Aldrich® vacuum filtration assembly
for 47 mm filters with glass support, NS 40/35 joints, 1 L flask (included)
Synonym(s):
Filtration Assembly, Vacuum Filtration System
All Photos(2)
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
Recommended Products
material
glass housing
glass membrane
Joint
joint ST/NS40/35
feature
graduation
packaging
pkg of 1 ea
bottle capacity
1 L
funnel capacity
300 mL
Related Categories
General description
Same as Z290432 but with NS 40/35 joint (male on flask, female on funnel support) instead of rubber stopper.
Vacuum filtration apparatus for HPLC solvent purification, particulate analysis, bacteriological studies, radiochemical determinations, quantitative gravimetric analysis, and similar applications. Clamp seals filter membrane between flanges on funnel top and funnel stem.
Includes: Funnel top, funnel stem/membrane support, aluminum clamp, stopper (except standard taper units) and filtration flask (where indicated).
For replacement parts, visit our online glassware catalog at www.sigma-aldrich.com
Includes: Funnel top, funnel stem/membrane support, aluminum clamp, stopper (except standard taper units) and filtration flask (where indicated).
For replacement parts, visit our online glassware catalog at www.sigma-aldrich.com
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
replacement part
Choose from one of the most recent versions:
Certificates of Analysis (COA)