SAB5201916
Monoclonal Anti-ERp57-Atto 488 antibody produced in mouse
clone 4F9, affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-PDIA3
About This Item
Recommended Products
biological source
mouse
conjugate
Atto 488 conjugate
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
4F9
monoclonal
description
clone 4F9, purified immunoglobulin, buffered aqueous solution
target molecular weight: 57 kDa
form
buffered aqueous solution
species reactivity
human (Tested)
packaging
pkg of 100 μg
storage condition
avoid repeated freeze/thaw cycles
protect from light
concentration
1.0 mg/mL
technique(s)
immunofluorescence: 1:100
western blot: 1:1000
isotype
IgG2b
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PDIA3(2923)
Immunogen
Application
Western blotting
Dilution 1:1000
Tissue/Cell Type Used: HeLa cell lysate
Immunofluorescence
Dilution 1:100
Tissue/Cell Type Used: HeLa
Physical form
Preparation Note
Storage and Stability
Other Notes
Immunogen region: Amino acids 25-505
Lot Expiration/Retest Date: Expires 1 year from purchase
Purification method: Protein G purified
Product type: Primary antibody
Antibody structure: Whole Antibody
Protein ID: NP_005304.3
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service