Skip to Content
Merck
All Photos(1)

Key Documents

74884

Supelco

Sodium 1-octanesulfonate monohydrate

≥99.0% (T)

Synonym(s):

1-Octanesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH2SO3Na · H2O
CAS Number:
Molecular Weight:
234.29
Beilstein:
3574241
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

anionic

Quality Level

Assay

≥99.0% (T)

form

sheet

mol wt

234.29 g/mol

technique(s)

ion pair chromatography: suitable

mp

≥300 °C (lit.)

SMILES string

O.[Na+].CCCCCCCCS([O-])(=O)=O

InChI

1S/C8H18O3S.Na.H2O/c1-2-3-4-5-6-7-8-12(9,10)11;;/h2-8H2,1H3,(H,9,10,11);;1H2/q;+1;/p-1

InChI key

MBURIAHQXJQKRE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 1-octanesulfonate monohydrate (NaOS) is an ion-pairing reagent.

Application

NaOS has been used as ion-pairing reagent for ultra-high performance liquid chromatography (UPLC) analysis with photodiode array detection, to estimate paraquat (methyl viologen) in tissue homogenate. It may also be used in determination of dopamine and its metabolites in brain microdialysates using liquid chromatography combined with electrospray ionization tandem mass spectrometry (LC–ESI-MS/MS).

Other Notes

Anionic detergent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Treves et al.
Canadian journal of biochemistry and cell biology = Revue canadienne de biochimie et biologie cellulaire, 62(1), 55-59 (1984-01-01)
Eight highly purified and crystalline enzymes representative of fundamental metabolic pathways and of cellular structure were tested with seven detergents. The enzyme-detergent interaction was studied as a time-independent (inhibition) or time-dependent (inactivation) phenomenon. Our results imply a specificity of detergent
Kamila Syslová et al.
Journal of chromatography. A, 1218(21), 3382-3391 (2011-03-15)
A sensitive assay method was developed for a parallel, rapid and precise determination of dopamine and its metabolites, homovanillic acid, 3-methoxytyramine and 3,4-dihydroxyphenylacetic acid, from brain microdialysates. The method consisted of a pre-treatment step, freeze-drying (lyophilization), to concentrate dopamine and
Rapid and simple analysis of paraquat in tissue homogenate by ultra-high performance liquid chromatography.
Merritt, T. J. S., et al.
Analytical Methods : Advancing Methods and Applications, 3.6, 1428-1432 (2011)
Maria Addolorata Saracino et al.
Journal of pharmaceutical and biomedical analysis, 104, 122-129 (2014-12-17)
A new microextraction by packed sorbent (MEPS) procedure coupled to a high-performance liquid chromatographic method has been developed for quantitation of catecholamines (i.e. norepinephrine, epinephrine and dopamine) in innovative biological samples, namely dried plasma and urine spots. Analyses were carried

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service