Skip to Content
Merck
Sodium 1-octanesulfonate monohydrate ≥99.0% (T)

Sodium 1-octanesulfonate monohydrate

Sodium 1-octanesulfonate monohydrate ≥99.0% (T)
Synonym(s):
1-Octanesulfonic acid sodium salt monohydrate
Linear Formula:
CH3(CH2)6CH2SO3Na · H2O
CAS Number:
Molecular Weight:
234.29
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Technique