Skip to Content
Merck
All Photos(1)

Key Documents

Z526207

Glas-Col® Mantleminder II

AC/DC input 240 V AC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112303
NACRES:
NB.76

manufacturer/tradename

Glas-Col 104A PL524

temperature measuring range

0-750 °C

L

6 in.

Looking for similar products? Visit Product Comparison Guide

General description

  • Time proportioning automatic temperature control
  • Type J immersion thermocouple included
  • Set-point in °C

Legal Information

Glas-Col is a registered trademark of Glas-Col LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service