Z247642
Coors™ high-alumina crucible
High form, capacity 100 mL
Synonym(s):
High-Alumina Crucible
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Coorstek 65505
H
61 mm
capacity
100 mL
top diam.
58 mm
Looking for similar products? Visit Product Comparison Guide
General description
Made of 99.8% pure, recrystallized aluminum oxide for high resistance to chemical attack. For use in reducing and oxidizing atmospheres and refractory metals (such as molybdenum, platinum, rhodium, tungsten, tantalum, and iridium). O.D. dimension tolerance is +/-5%. Max. use temp. (no load): 1,750 °C.
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service