SIK1103-10
12,12,13,13,14,14,15,15,15-Nonafluoropentadecane-1-thiol
97%
About This Item
Recommended Products
Assay
97%
form
solid
shelf life
3 years under the recommended conditions
color
white
SMILES string
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)CCCCCCCCCCCS
InChI
1S/C15H23F9S/c16-12(17,13(18,19)14(20,21)15(22,23)24)10-8-6-4-2-1-3-5-7-9-11-25/h25H,1-11H2
InChI key
QYLOPCNGRGFDFY-UHFFFAOYSA-N
Related Categories
Application
The fluorinated carbon gives omniphobic properties to the surface. The coated surface is more water repellent when a product having more fluorinated carbons is used.
The thiol group provide a linker that covalently bond to the coinage metal surface.
The long carbon chain length allows for easy formation of a self-assembled monolayer.
Legal Information
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service