Skip to Content
Merck

SIK1101-10

Sigma-Aldrich

12,12,13,13,14,14,15,15,16,16,17,17,18,18,19,19,19-Heptadecafluorononadecanethiol

97%

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C19H23F17S
CAS Number:
Molecular Weight:
606.42
UNSPSC Code:
12352105
NACRES:
NA.23
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Assay

97%

form

solid

shelf life

3 years under the recommended conditions

color

white

Application

These products are used to form water-repellent surface coating and/or lubricated surfaces on coinage metal materials (such as gold, silver).
The fluorinated carbon gives omniphobic properties to the surface. The coated surface is more water repellent when a product having more fluorinated carbons is used.
The thiol group provide a linker that covalently bond to the coinage metal surface.
The long carbon chain length allows for easy formation of a self-assembled monolayer.

Legal Information

Product of SiKEMIA

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service