DWK218667339
Duran® Laboratory Bottle Wide-Neck Amber
amber bottle, protect, without cap, GLS 80 (without ring), capacity 5000 mL
Synonym(s):
218667339, Amber Lab Bottle, Wide-Neck Amber Bottle
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
amber bottle
feature
without cap GLS 80 (without ring)
packaging
pkg of 1 ea
manufacturer/tradename
DURAN® 218667339
capacity
5000 mL
Looking for similar products? Visit Product Comparison Guide
Application
Preparation of light-sensitive materials, Storage of light-sensitive materials, Transport and shipping of light-sensitive materials,Boro 3.3, plastic coated
Features and Benefits
Easy access to the bottle contents due to the wide diameter of the bottle neck ; Optimum filling and pouring and particularly easy cleaning; Spoons, spatulas and tweezers can be manipulated with ease; Even large-volume funnels can be inserted into the bottles without any difficulty ; Stability due to the large base area; Easy-to-read scale with graduation marks to estimate the content; Fired-on, highly durable white ceramic; UV protection up to approx. 500 nm due to external colouration; The only external application of the colour ensures that internal surfaces have standard DURAN® glass properties;
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service