DWK218010851
DURAN® graduated laboratory bottle
with cap, round glass bottle, bottle capacity (10 mL), non-sterile
Synonym(s):
218010851
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
round glass bottle
sterility
non-sterile
feature
with cap
packaging
case of 10 ea
manufacturer/tradename
DURAN® 218010851
bottle capacity
(10 mL)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
with DIN 168-1 Thread, graduated
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service