424560
1-[Bis[3-(dimethylamino)propyl]amino]-2-propanol
98%
Synonym(s):
N ,N -Bis(3-dimethylaminopropyl)-N -(2-hydroxypropyl)amine
About This Item
Recommended Products
Assay
98%
refractive index
n20/D 1.459 (lit.)
bp
290 °C (lit.)
density
0.89 g/mL at 25 °C (lit.)
SMILES string
CC(O)CN(CCCN(C)C)CCCN(C)C
InChI
1S/C13H31N3O/c1-13(17)12-16(10-6-8-14(2)3)11-7-9-15(4)5/h13,17H,6-12H2,1-5H3
InChI key
NWDRKFORNVPWLY-UHFFFAOYSA-N
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service