282804
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane
98%
Synonym(s):
1,4,8,11-Tetramethylcyclam
About This Item
Recommended Products
Quality Level
Assay
98%
form
solid
mp
38-42 °C (lit.)
SMILES string
CN1CCCN(C)CCN(C)CCCN(C)CC1
InChI
1S/C14H32N4/c1-15-7-5-8-17(3)13-14-18(4)10-6-9-16(2)12-11-15/h5-14H2,1-4H3
InChI key
HRFJEOWVAGSJNW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.
Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.
Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.
Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.
Protocols
We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.
We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.
Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.
Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service