Skip to Content
Merck
All Photos(2)

Key Documents

372900

Sigma-Aldrich

3-Methyl-1,1-diphenylurea

≥95.0%

Synonym(s):

N-Methyl-N′,N′-diphenylurea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2NCONHCH3
CAS Number:
Molecular Weight:
226.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0%

form

solid

mp

172-174 °C (lit.)

functional group

amine

SMILES string

CNC(=O)N(c1ccccc1)c2ccccc2

InChI

1S/C14H14N2O/c1-15-14(17)16(12-8-4-2-5-9-12)13-10-6-3-7-11-13/h2-11H,1H3,(H,15,17)

InChI key

IMFYAZJNDOZIFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-1,1-diphenylurea is reported as USP standard.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - STOT RE 2 Oral

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Łukasz Jedynak et al.
Journal of pharmaceutical and biomedical analysis, 83, 19-27 (2013-05-25)
An HPLC method for determination of related substances in temozolomide drug substance was developed. Particular attention was paid to the stability studies due to the fact that temozolomide is unstable in a solution and quickly decomposes to its main degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service