Skip to Content
Merck
3-Methyl-1,1-diphenylurea ≥95.0%

3-Methyl-1,1-diphenylurea

3-Methyl-1,1-diphenylurea ≥95.0%
Synonym(s):
N-Methyl-N′,N′-diphenylurea
Linear Formula:
(C6H5)2NCONHCH3
CAS Number:
Molecular Weight:
226.27
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing