254673
1,3,5-Trimethylhexahydro-1,3,5-triazine
97%
Synonym(s):
Hexahydro-1,3,5-trimethyl-1,3,5-triazine
About This Item
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.4621 (lit.)
bp
162-163.5 °C (lit.)
mp
−27 °C (lit.)
density
0.919 g/mL at 25 °C (lit.)
SMILES string
CN1CN(C)CN(C)C1
InChI
1S/C6H15N3/c1-7-4-8(2)6-9(3)5-7/h4-6H2,1-3H3
InChI key
DPMZXMBOYHBELT-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
123.8 °F - closed cup
Flash Point(C)
51 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service