224413
Cyclohexylmagnesium chloride solution
2.0 M in diethyl ether
Synonym(s):
Chlorocyclohexylmagnesium
About This Item
Recommended Products
reaction suitability
reaction type: Grignard Reaction
concentration
2.0 M in diethyl ether
density
0.871 g/mL at 25 °C
SMILES string
Cl[Mg]C1CCCCC1
InChI
1S/C6H11.ClH.Mg/c1-2-4-6-5-3-1;;/h1H,2-6H2;1H;/q;;+1/p-1
InChI key
WMJMABVHDMRMJA-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-40.0 °F - closed cup
Flash Point(C)
-40 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service