Skip to Content
Merck
All Photos(1)

Key Documents

224405

Sigma-Aldrich

Cyclopentylmagnesium chloride solution

2.0 M in diethyl ether

Synonym(s):

Chloro cyclopentyl magnesium, Chlorocyclopentylmagnesium, Cyclopentyl magnesium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9ClMg
CAS Number:
Molecular Weight:
128.88
Beilstein:
4124474
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in diethyl ether

density

0.878 g/mL at 25 °C

SMILES string

Cl[Mg]C1CCCC1

InChI

1S/C5H9.ClH.Mg/c1-2-4-5-3-1;;/h1H,2-5H2;1H;/q;;+1/p-1

InChI key

GROGEABZCKQTJW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleg V Larionov et al.
Organic letters, 16(3), 864-867 (2014-01-15)
A one-step transformation of heterocyclic N-oxides to 2-alkyl-, aryl-, and alkenyl-substituted N-heterocycles is described. The success of this broad-scope methodology hinges on the combination of copper catalysis and activation by lithium fluoride or magnesium chloride. The utility of this method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service