Skip to Content
Merck
All Photos(1)

Key Documents

DLD-1

90102540, human colon, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

DLD-1, 90102540

biological source

human colon

growth mode

Adherent

karyotype

2n = 46, pseudodiploid

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon adenocarcinoma

Cell Line Description

Derived from human colorectal adenocarcinoma. The cells have been used in the study of polar solvents on cell characteristics.The four cell lines: DLD-1 (90102540), HCT-15 (91030712), HCT-8 (90032006) and HRT-18 (86040306) have been shown to have a common genetic origin.

Application

DLD-1 cell line has been used to express shRNA oligos directed against coronin 2A (CRN5) and to study the role of CRN5 in tumor cell migration. It has also been used to determine the efficiency of antibody-coupled short interfering RNA (siRNA) for in vivo mRNA knockdown.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 8,11
D16S539: 12,13
D5S818: 13
D7S820: 10,12
THO1: 7,9.3
TPOX: 8,11
vWA: 18,19

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 1-3 x 10,000 cells/cm2 using 0.25% trypsin, 0.02% EDTA.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service