AGS Cell Line human
89090402, human stomach, Not specified
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
product name
AGS Cell Line human, 89090402
biological source
human stomach
description
Human Caucasian gastric adenocarcinoma
growth mode
Adherent
karyotype
Hyperdiploid
morphology
Not specified
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian gastric adenocarcinoma
Cell Line Description
Derived from an adenocarcinoma of the stomach of a 54 year-old Caucasian female with no prior anti-cancer treatment. AGS cells are persistently infected with parainfluenza virus type 5 (PIV5).
Application
AGS human cell line has been used:
- to study the role of obestatin/G protein-coupled receptor 39 (GPR39) in gastric cancer progression
- to determine expression levels of colon cancer associated transcript -1 (CCAT1)
- to study the adhesion of Lactobacillus sps. to the gastric mucosa
Anti-tumour testing
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 12
D16S539: 11,13
D5S818: 9,12
D7S820: 10,11
THO1: 6,7
TPOX: 11,12
vWA: 16,17
CSF1PO: 11,12
D13S317: 12
D16S539: 11,13
D5S818: 9,12
D7S820: 10,11
THO1: 6,7
TPOX: 11,12
vWA: 16,17
Culture Medium
Ham′s F12 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin; 0.05% EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service