Skip to Content
Merck
All Photos(1)

Key Documents

05-4600

Sigma-Aldrich

Chromium(VI) oxide

SAJ special grade, ≥98.0%

Synonym(s):

Chromic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CrO3
CAS Number:
Molecular Weight:
99.99
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:

grade

SAJ special grade

Assay

≥98.0%

form

solid

availability

available only in Japan

pH

<1 (20 °C, 100 g/L)

mp

196 °C (dec.) (lit.)

SMILES string

O=[Cr](=O)=O

InChI

1S/Cr.3O

InChI key

WGLPBDUCMAPZCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kondo et al.
European journal of biochemistry, 200(3), 689-698 (1991-09-15)
The chemical structure of the saccharide portion of Vibrio parahaemolyticus serotype 012 lipopolysaccharide was studied. Using chemical degradation and modification, as well as methylation analysis in combination with GLC-MS, laser-desorption mass spectrometry and 1H-NMR and 13C-NMR spectroscopy, the carbohydrate backbone
Seongmin Lee et al.
Organic letters, 11(1), 5-8 (2008-12-06)
The C14,15-dihydro-C22,25-epi north unit of cephalostatin 1 has been synthesized in 11 operations from commercially available hecogenin acetate via multiple reductions and oxidations. The key transformations include (i) Cr(VI)-catalyzed E-ring opening, (ii) C17 hydroxylation, and (iii) a base-triggered cyclization cascade.
M C García-Rodríguez et al.
Mutation research, 496(1-2), 145-151 (2001-09-12)
The effect of chlorophyllin on micronucleated polychromatic erythrocytes (MN-PCE) induction by chromium trioxide (CrO(3)) exposure in peripheral blood of mice was studied. Animals were treated with a single intraperitoneal dose of chlorophyllin (CHL) (20mg/kg), CrO(3) (20mg/kg), and CHL (20mg/kg) 4h
L Vayssieres et al.
Journal of nanoscience and nanotechnology, 1(4), 385-388 (2003-08-14)
We are reporting here on the inexpensive fabrication and optical properties of an iron(III) oxide-chromium(III) oxide nanocomposite thin film of corundum crystal structure. Its novel and unique-designed architecture consists of uniformed, well-defined and oriented nanorods of Hematite (alpha-Fe2O3) of 50
Adam Szelag et al.
Polish journal of pharmacology, 55(6), 1097-1103 (2004-01-20)
Hexavalent chromium compounds exhibit higher toxicity than its trivalent compounds since chromium ions in the +6 oxidation state easily cross biological membranes. It has recently been proposed that substances reducing chromium ions from the +6 to the less toxic +3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service