Merck
  • Chemical structure of the carbohydrate backbone of Vibrio parahaemolyticus serotype 012 lipopolysaccharide.
Chemical structure of the carbohydrate backbone of Vibrio parahaemolyticus serotype 012 lipopolysaccharide.
European journal of biochemistry (1991-09-15)
S Kondo, U Zähringer, U Seydel, V Sinnwell, K Hisatsune, E T Rietschel
ABSTRACT
The chemical structure of the saccharide portion of Vibrio parahaemolyticus serotype 012 lipopolysaccharide was studied. Using chemical degradation and modification, as well as methylation analysis in combination with GLC-MS, laser-desorption mass spectrometry and 1H-NMR and 13C-NMR spectroscopy, the carbohydrate backbone of the lipopolysaccharide was characterized as a branched decasaccharide with the following structure: (formula; see text) In the native lipopolysaccharide two additional phosphate groups are present and 3-deoxy-D-threo-hexulosonic acid and D-galacturonic acid are bound via acid-labile linkages.
Sigma-Aldrich
Chromium(VI) oxide, SAJ special grade, ≥98.0%
Sigma-Aldrich
Chromium(VI) oxide, 99.99% trace metals basis
Sigma-Aldrich
Chromium(VI) oxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Chromium(VI) oxide, ReagentPlus®, 99.9% trace metals basis