Skip to Content
Merck
All Photos(2)

Key Documents

5.08578

Sigma-Aldrich

TSPO Agonist, Ro5-4864

Synonym(s):

TSPO Agonist, Ro5-4864, 4ʹ-chlorodiazepam, TSPO Activator I, Mitochondrial Translocator Protein Activator I, Ro54864

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12Cl2N2O
CAS Number:
Molecular Weight:
319.19
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

4.4 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)C2=NCC(=O)N(c3c2cc(cc3)Cl)C

InChI

1S/C16H12Cl2N2O/c1-20-14-7-6-12(18)8-13(14)16(19-9-15(20)21)10-2-4-11(17)5-3-10/h2-8H,9H2,1H3

InChI key

PUMYFTJOWAJIKF-UHFFFAOYSA-N

General description

A 4′-chloro derivative of diazepam that lacks affinity for GABA-A receptors, but can serve as a ligand for the peripheral benzodiazepine receptor. Acts as an agonist for the 18 kDa translocator protein (TSPO) on the outer mitochondrial membrane (Kd = 4.4 nM). Diminishes the accumulation of β-amyloid peptides in the CA1 hippocampal region of gonadectomized (GDX) mice and in triple transgenic Alzheimer′s disease murine models (3xTgAD). However, it does not affect tau phosphorylation events as assessed by AT8 immunoreactivity. Also shown to reduce GFAP ionized calcium binding adaptor molecule-1 (IBA-1) in the 3xTgAD mice. Reported to increase testosterone and progesterone levels in GDX mice without affecting other steroid hormones. However, it does not have a significant effect on the levels of these hormones in aged 3xTgAD mice. In combination with another mitochondria targeting molecule, lonidamine, it is shown to induce apoptosis in MDA-MB-231 breast cancer cells.
A 4′-chloro derivative of diazepam that lacks affinity for GABA-A receptors, but can serve as a ligand for the peripheral benzodiazepine receptor. Acts as an agonist for the 18 kDa translocator protein (TSPO) on the outer mitochondrial membrane (Kd = 4.4 nM). Diminishes the accumulation of β-amyloid peptides in the CA1 hippocampal region of gonadectomized (GDX) mice and in triple transgenic Alzheimer′s disease murine models (3xTgAD). However, it does not affect tau phosphorylation events as assessed by AT8 immunoreactivity. Also shown to reduce GFAP ionized calcium binding adaptor molecule-1 (IBA-1) in the 3xTgAD mice. Reported to increase testosterone and progesterone levels in GDX mice without affecting other steroid hormones. However, it does not have a significant effect on the levels of these hormones in aged 3xTgAD mice. In combination with another mitochondria targeting molecule, lonidamine, it is shown to induce apoptosis in MDA-MB-231 breast cancer cells.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TSPO

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Barron, A.M., et al. 2013. The J.Neuroscience.33, 8891.Yousefi, O.S., et al. 2013. Cell Commun.Signaling.11, 13.Wu, X., and Gallo, K.E. 2013. PLoS One.8, e71258.Leonelli, E., et al. 2005. Mech. Ageing. Dev.126, 1159.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service