01-107
Epidermal growth factor, human
suitable for cell culture
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32
Recommended Products
General description
FUNCTION: SwissProt: P01133 # EGF stimulates the growth of various epidermal and epithelial tissues in vivo and in vitro and of some fibroblasts in cell culture.
SIZE: 1207 amino acids; 133946 Da
SUBCELLULAR LOCATION: Membrane; Single-pass type I membrane protein.
SIMILARITY: SwissProt: P01133 ## Contains 9 EGF-like domains. & Contains 9 LDL-receptor class B repeats.
SIZE: 1207 amino acids; 133946 Da
SUBCELLULAR LOCATION: Membrane; Single-pass type I membrane protein.
SIMILARITY: SwissProt: P01133 ## Contains 9 EGF-like domains. & Contains 9 LDL-receptor class B repeats.
Quality
Routinely evaluated in a 5-day growth assay using human foreskin fibroblasts
Physical form
Lyophilized from 100mM acetic acid with 3.5mg mannitol
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service