Skip to Content
Merck

01-407

Epidermal growth factor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
eCl@ss:
32160405
Biological source:
human
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Epidermal growth factor (EGF),

biological source

human

Quality Segment

description

Neural Stem Cells

purified by

filtration through 0.2μm medium

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

Gene Information

human ... EGF(1950)

General description

FUNCTION: SwissProt: P01133 # EGF stimulates the growth of various epidermal and epithelial tissues in vivo and in vitro and of some fibroblasts in cell culture.
SIZE: 1207 amino acids; 133946 Da
SUBCELLULAR LOCATION: Membrane; Single-pass type I membrane protein.
SIMILARITY: SwissProt: P01133 ## Contains 9 EGF-like domains. & Contains 9 LDL-receptor class B repeats.

Physical form

Lypholized from 100mM acetic acid with 3.5mg mannitol

Analysis Note

routinely evaluated in a 5-day growth assay using human foreskin fibroblasts

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library