Skip to Content
Merck
All Photos(1)

Key Documents

900621

Sigma-Aldrich

CyJohnPhos Pd G3

95%

Synonym(s):

(2-Dicyclohexylphosphino-1,1′-biphenyl)[2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate, CyJohnPhos G3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H44NO3PPdS
CAS Number:
Molecular Weight:
720.21
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

95%

form

powder or crystals

feature

generation 3

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

SMILES string

O=S(=O)([O-][Pd+2]1([C-]=2C=CC=CC2C=3C=CC=CC3[NH2]1)[P](C=4C=CC=CC4C=5C=CC=CC5)(C6CCCCC6)C7CCCCC7)C

InChI

InChI=1S/C24H31P.C12H10N.CH4O3S.Pd/c1-4-12-20(13-5-1)23-18-10-11-19-24(23)25(21-14-6-2-7-15-21)22-16-8-3-9-17-22;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h1,4-5,10-13,18-19,21-22H,2-3,6-9,14-17H2;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;-1;;+2/p-1

InChI key

NFZUZGPTBAUUOC-UHFFFAOYSA-M

Application

Third generation (G3) Buchwald palladium precatalyst for cross-coupling, demonstrating improved stability and reactivity from previous generations. This same product was formerly listed under RNI00192.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service