Skip to Content
Merck
All Photos(1)

Key Documents

818393

TSKgel® GMHHR-H(S) HT (13 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, polystyrene-divinylbenzene phase

Synonym(s):

TSKgel® GPC Columns for Applications up to 140° C

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
20.740,00 kr.

20.740,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
20.740,00 kr.

About This Item

UNSPSC Code:
41115711

20.740,00 kr.


Please contact Customer Service for Availability

Product Name

TSKgel® GMHHR-H(S) HT HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 13 μm particle size

material

stainless steel hardware

Agency

suitable for USP L21

product line

TSKgel®

manufacturer/tradename

Tosoh 18393

parameter

140 °C temperature
2.5 mL/min flow rate
20 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

polymer particle platform

matrix active group

polystyrene-divinylbenzene phase

particle size

13 μm

pore size

mixed

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel HT (High Temperature) columns are for the analysis of organic-soluble polymers at temperatures up to 140°C and are shipped in o-dichlorobenzene (ODCB), which can be exchanged for 1-chloronaphthalene and trichlorobenzene. The TSKgel mixed bed columns are packed with particles of different pore sizes that are blended to provide an extended linear calibration curve. Mixed bed columns are used when a broad range of polymer molar masses is expected in the sample. Shearing, which can be observed when analyzing ultra-high molar mass polymers on columns packed with 5 micron or smaller particles, can be overcome by using the TSKgel GMHhr-H(S) HT column, which is packed with 13 micron particles. The (S) is a reference to this shearing effect.
The issue of shearing that occurs with the analysis of ultra-high molar mass polymers is overcome by the TSKgel GMHhr-H(S) HT column. The (S) is a reference to this shearing effect.

Physical form

ODCB can be exchanged by 1-chloronaphthalene and trichlorobenzene.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service