![TSKgel® GMHHR-H(S) HT HPLC Column phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 13 μm particle size](/deepweb/assets/sigmaaldrich/product/images/334/931/848b78a7-70aa-4507-9d11-5941c9b0fa1f/640/848b78a7-70aa-4507-9d11-5941c9b0fa1f.jpg)
TSKgel® GMHHR-H(S) HT (13 µm) HPLC Columns
![TSKgel® GMHHR-H(S) HT HPLC Column phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 13 μm particle size](/deepweb/assets/sigmaaldrich/product/images/334/931/848b78a7-70aa-4507-9d11-5941c9b0fa1f/640/848b78a7-70aa-4507-9d11-5941c9b0fa1f.jpg)
Synonym(s):
TSKgel® GPC Columns for Applications up to 140° C
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing