Skip to Content
Merck
All Photos(1)

Key Documents

28947-U

Supelco

Tenax® TA

stainless steel TD tube, unconditioned

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104000

material

stainless steel TD tube

Agency

EPA TO-1,IP-1B

feature

unconditioned

packaging

pkg of 10 ea

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Tenax TA

application(s)

environmental

compatibility

for use with PerkinElmer ATD-50,ATD-400 and TurboMatrix (TMX) Series

Looking for similar products? Visit Product Comparison Guide

Legal Information

Tenax is a registered trademark of Buchem B.V.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service