Skip to Content
Merck
All Photos(4)

Key Documents

20010-U

Supelco

Tenax® TA Thermal Desorption Tube

greener alternative

stainless steel, O.D. x I.D. x L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea

Synonym(s):

Tenax TA Thermal Desorption Tube (stainless steel)

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
10.100,00 kr.

10.100,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 EA
10.100,00 kr.

About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

10.100,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Tenax® TA, stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, 60-80 mesh, pkg of 10 ea

material

stainless steel TD tube

Quality Level

Agency

EPA TO-1,IP-1B

description

Sealed with TDS³ Storage Container

feature

preconditioned

packaging

pkg of 10 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Tenax TA porous polymer

particle size

60-80 mesh

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Tenax® TA is a preconditioned, stainless steel thermal desorption tube.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Application


  • Determination of emission factors from a landfill through an inverse methodology: Experimental determination of ambient air concentrations and use of numerical modelling.: This study focuses on identifying emission factors from landfills using a combination of experimental data collection of ambient air and numerical modeling to better understand environmental impacts (Gallego et al., 2024 Apr 28) (Gallego et al., 2024).

  • Analysis of volatile organic compounds in Korean-bred strawberries: insights for improving fruit flavor.: This research analyzes the profiles of volatile organic compounds in strawberries to enhance their flavor, providing insights valuable for agricultural and food chemistry sectors (Jee et al., 2024) (Jee et al., 2024).

  • Bioaccessibility of halogenated flame retardants and organophosphate esters in settled dust: Influences of specific dust matrices from informal e-waste and end-of-life vehicle processing areas in Vietnam.: This study evaluates the bioaccessibility of various flame retardants in dust samples from e-waste recycling areas, contributing to environmental health and safety knowledge (Hoang et al., 2024 May 20) (Hoang et al., 2024).

  • Effect of spraying air freshener on particulate and volatile organic compounds in vehicles.: Investigating the impact of air fresheners on the air quality inside vehicles, this study provides insights into the changes in particulate and volatile organic compound concentrations, relevant for environmental health (Lee et al., 2024 Mar 15) (Lee et al., 2024).

  • A Compact Monitor for Ethylene and Other Plant-Produced Volatile Organic Compounds for NASA Space Missions.: This article describes the development of a compact monitoring system for ethylene and other VOCs in a space environment, highlighting its applications in space biology and physical sciences (Dobrokhotov et al., 2023 Dec 8) (Dobrokhotov et al., 2023).


Legal Information

Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hermann Fromme et al.
Environment international, 126, 145-152 (2019-02-25)
Linear and cyclic volatile methylsiloxanes (l-VMS and c-VMS) are man-made chemicals with no natural source. They have been widely used in cosmetics, personal care products, coatings and many other products. As a consequence of their wide use, VMS can be
Wolfgang Schober et al.
International journal of hygiene and environmental health, 222(3), 486-493 (2019-01-28)
Smoking in car interiors is of particular concern because concentrations of potentially harmful substances can be expected to be high in such small spaces. To assess the potential exposure for occupants, especially children, we performed a comprehensive evaluation of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service