Documents
21910
SGE Syringes for Agilent/HP 7673, 7683 and 6850 ALS Autosampler, Fixed Needle
volume 5 μL, SK-5F-HP-0.47, needle size 26 ga, needle L 42 mm, pkg of 6 ea
About This Item
UNSPSC Code:
41122001
material
cone tip stainless steel needle (point style 1)
needle size
26 ga
feature
needle type F (fixed)
packaging
pkg of 6 ea
manufacturer/tradename
SGE 001804
needle L
42 mm
volume
5 μL
compatibility
for use with Agilent/HP 7673 and 7683 autosam pler
Related Categories
General description
SGE syringes are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most troublefree autosampler syringes available.
Choose from one of the most recent versions:
Certificates of Analysis (COA)