Key Documents
21544
SGE Syringes for Agilent/HP 7673, 7683 and 6850 ALS Autosampler, Fixed Needle
volume 10 μL, SK-10F-HP-0.63, needle size 23 ga, needle L 42 mm, pkg of 6 ea
Select a Size
6 EA
1.320,00 kr.
1.320,00 kr.
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
6 EA
1.320,00 kr.
About This Item
UNSPSC Code:
41122001
1.320,00 kr.
Please contact Customer Service for Availability
Request a Bulk Order
material
cone tip stainless steel needle (point style 1)
needle size
23 ga
feature
needle type F (cemented needle)
packaging
pkg of 6 ea
manufacturer/tradename
SGE 002814
needle L
42 mm
volume
10 μL
compatibility
for use with Agilent/HP 7673 and 7683 autosampler
General description
SGE syringes are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most troublefree autosampler syringes available.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Krzysztof Kotowski et al.
Anticancer research, 40(5), 2613-2625 (2020-05-06)
The occurrence of BRAFV600E mutation causes an up-regulation of the B-raf kinase activity leading to the stabilization of hypoxia-inducible factor 1-alpha (HIF-1α) - the promoter of the 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3 (PFKFB3) enzyme. The aim of the study was to examine the