Key Documents
Z744145
Wilmad® 50 ml Suprasil® Small Volume Liquid Nitrogen Dewar
offset for Bruker®, pack of 1
Synonym(s):
EPR cryo dewar, EPR dewar
About This Item
UNSPSC Code:
41121804
packaging
pack of 1
manufacturer/tradename
Wilmad WG-819-B-Q
General description
Dewar Flasks provide a convenient alternative to Variable Temperature systems for studies at 77 K, and are silvered around the reservoir. The offset versions (WG-819) are designed for cavities that have limited space between the reservoir and the drive shaft for iris.
Legal Information
Bruker is a registered trademark of Bruker-Physik AG
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)