Skip to Content
Merck
All Photos(1)

Key Documents

SML3408

Sigma-Aldrich

EG00229 trifluoroacetate

≥98% (HPLC)

Synonym(s):

N2-​[[3-​[(2,​1,​3-​Benzothiadiazol-​4-​ylsulfonyl)​amino]​-​2-​thienyl]​carbonyl]​-L-​arginine trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
884,00 kr.
25 MG
3.550,00 kr.

884,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
884,00 kr.
25 MG
3.550,00 kr.

About This Item

Empirical Formula (Hill Notation):
C17H19N7O5S3·CF3COOH
CAS Number:
Molecular Weight:
611.60
UNSPSC Code:
51111800
NACRES:
NA.77

884,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

storage temp.

−20°C

SMILES string

FC(F)(F)C(=O)O.[S](=O)(=O)(Nc3c([s]cc3)C(=O)N[C@@H](CCCN=C(N)N)C(=O)O)c1c2n[s]nc2ccc1

InChI

1S/C17H19N7O5S3.C2HF3O2/c18-17(19)20-7-2-4-11(16(26)27)21-15(25)14-10(6-8-30-14)24-32(28,29)12-5-1-3-9-13(12)23-31-22-9;3-2(4,5)1(6)7/h1,3,5-6,8,11,24H,2,4,7H2,(H,21,25)(H,26,27)(H4,18,19,20);(H,6,7)/t11-;/m0./s1

InChI key

ZYQBITUOSRZDTG-MERQFXBCSA-N

Biochem/physiol Actions

Antagonist of the VEGF-A receptor neuropilin 1 (NRP1)
EG00229 trifluoroacetate is an antagonist of the VEGF-A receptor neuropilin 1 (NRP1), which decreases viability of A549 lung carcinoma cells. EG00229 trifluoroacetate impairs VEGFA induced migration of HUV-EC-C endothelial cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James L Daly et al.
Science (New York, N.Y.), 370(6518), 861-865 (2020-10-22)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), the causative agent of coronavirus disease 2019 (COVID-19), uses the viral spike (S) protein for host cell attachment and entry. The host protease furin cleaves the full-length precursor S glycoprotein into two associated
Ashley Jarvis et al.
Journal of medicinal chemistry, 53(5), 2215-2226 (2010-02-16)
We report the molecular design and synthesis of EG00229, 2, the first small molecule ligand for the VEGF-A receptor neuropilin 1 (NRP1) and the structural characterization of NRP1-ligand complexes by NMR spectroscopy and X-ray crystallography. Mutagenesis studies localized VEGF-A binding
Jillian C Nissen et al.
Journal of neurochemistry, 127(3), 394-402 (2013-09-17)
Tuftsin (Thr-Lys-Pro-Arg) is a natural immunomodulating peptide found to stimulate phagocytosis in macrophages/microglia. Tuftsin binds to the receptor neuropilin-1 (Nrp1) on the surface of cells. Nrp1 is a single-pass transmembrane protein, but its intracellular C-terminal domain is too small to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service