Skip to Content
Merck
All Photos(1)

Documents

SCP0011

Sigma-Aldrich

Akt Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60N14O8
Molecular Weight:
816.95
UNSPSC Code:
12352200
NACRES:
NA.32

product name

Akt Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥65%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Arg-Pro-Arg-Ala-Ala-Thr-Phe-NH2

Application

AKT refers to the serine/threonine protein kinases, AKT-1, AKT-2 and AKT-3, which are involved in the cellular processes of glucose metabolism, cell proliferation, apoptosis, transcription and cell migration. The peptide Arg-Pro-Arg-Ala-Ala-Thr-Phe may be a relatively specific substrate for PKBα (AKT-1) because, unlike other substrates, it is not phosphorylated by p70 S6 kinase or MAP kinase activated protein (MAPKAP) kinase-1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John N Hutchinson et al.
Cancer research, 64(9), 3171-3178 (2004-05-06)
Elevated expression of Akt-1 (PKBalpha) has been noted in a significant percentage of primary human breast cancers. Another frequent event in the genesis of human breast cancers is amplification and overexpression of the ErbB-2 receptor tyrosine kinase, an event which
D R Alessi et al.
FEBS letters, 399(3), 333-338 (1996-12-16)
The substrate specificity of protein kinase-B alpha (PKBalpha, also known as RAC kinase or Akt) was investigated using synthetic peptide substrates related to the sequence surrounding the phosphorylation site on glycogen synthase kinase-3 (GSK3). The minimum sequence motif required for
G Rena et al.
The Journal of biological chemistry, 274(24), 17179-17183 (1999-06-08)
Protein kinase B lies "downstream" of phosphatidylinositide (PtdIns) 3-kinase and is thought to mediate many of the intracellular actions of insulin and other growth factors. Here we show that FKHR, a human homologue of the DAF16 transcription factor in Caenorhabditis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service