Skip to Content
Merck
All Photos(1)

Key Documents

SML1367

Sigma-Aldrich

EG00229

≥98% (HPLC)

Synonym(s):

N2-[[3-[(2,1,3-Benzothiadiazol-4-ylsulfonyl)amino]-2-thienyl]carbonyl]-L-arginine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N7O5S3
CAS Number:
Molecular Weight:
497.57
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

, pale yellow to yellow-orange

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N[C@H](C(O)=O)CCCNC(N)=N)C1=C(C=CS1)NS(C2=CC=CC3=NSN=C32)(=O)=O

InChI

1S/C17H19N7O5S3/c18-17(19)20-7-2-4-11(16(26)27)21-15(25)14-10(6-8-30-14)24-32(28,29)12-5-1-3-9-13(12)23-31-22-9/h1,3,5-6,8,11,24H,2,4,7H2,(H,21,25)(H,26,27)(H4,18,19,20)/t11-/m0/s1

InChI key

ZWWMEDURALZMEV-NSHDSACASA-N

Biochem/physiol Actions

EG00229 is an antagonist of the VEGF-A receptor neuropilin 1 (NRP1), which decreases viability of A549 lung carcinoma cells. EG00229 impairs VEGFA induced migration of HUV-EC-C endothelial cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service