Key Documents
MT30671569
Sigma-Aldrich
EasyPlace Electrode Arm
complete
Select a Size
1 EACH
2.450,00 kr.
2.450,00 kr.
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
2.450,00 kr.
About This Item
UNSPSC Code:
23242102
NACRES:
NB.79
2.450,00 kr.
Please contact Customer Service for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Mettler-Toledo 30671569
compatibility
for use with Mettler-Toledo meters
General description
Precise Control
Perfectly vertical sensor movement makes it easy to insert the sensor into samples.
The right place
The EasyPlace contains dedicated positions for calibration, short- and long term sensor storage and measurement, simplifying everyday workflows.
Fits right in
The EasyPlace electrode arm fits together with the EasyMix magnetic stirrer and standard sachets, simplifying operation and saving space.
Choose from one of the most recent versions:
Certificates of Analysis (COA)