Skip to Content
Merck
All Photos(1)

Key Documents

MT30671556

Sigma-Aldrich

Mettler Toledo SevenDirect SD20 pH / ORP Meter HA Kit

electrode, InLab® Routine Pro-ISM

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
13.350,00 kr.

13.350,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
13.350,00 kr.

About This Item

UNSPSC Code:
41115603
NACRES:
NB.61

13.350,00 kr.


Please contact Customer Service for Availability

AC/DC input

, universal plug set
220 V / 240 V

feature

electrode InLab® Routine Pro-ISM

packaging

pack of 1 kit

manufacturer/tradename

Mettler-Toledo 30671556

mV measuring range

-2,000.000-2,000.000 mV, resolution: 1; 0.1

pH measuring range

-2.000-20.000, accuracy: ±0.002, resolution: 0.1
resolution: 0.001
resolution: 0.01

temperature measuring range

-30-130 °C, resolution: 0.1 °C

General description

Mastered in no Time
Large color touchscreen provides direct measurement and easy data entry. Instructions guide users through the sensor calibration process.
Tracks your Records
The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
Tough Enough
Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
Chemically resistant pH Sensor
InLab Routine-Pro ISM is a refillable pH/temperature sensor for many applications made from chemically resistant glass. The HA membrane glass minimizes alkali errors and makes it especially suitable for measuring at high pH values.
Electrode Arm for Efficient Operation
The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy. Dedicated positions for cleaning and storage support correct sensor handling. Vertical and horizontal movements facilitate precise positioning, even with one hand.
Protect Critical Settings
A two-level user management protects the most important settings. Operators retain the flexibility to individually set preferences like language and screen brightness.
Display Adapts to Your Needs
View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.

Application

pH measurement

Features and Benefits

  • Large color touchscreen provides direct measurement and easy data entry.
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • Smart design and cover keeps the instrument safe from dust and spills
  • The HA membrane glass makes it especially suitable for measuring at high pH values.
  • Electrode arm for efficient operation
  • Protect critical settings Protected with two-level user management
  • Display adapts to your needs to maximize readability

Components

Includes: Meter;Sensor; an efficient electrode arm; power supply; pH buffer sachets for initial calibration.

Legal Information

InLab is a registered trademark of Mettler-Toledo, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service