Skip to Content
Merck
All Photos(1)

Documents

M5634

Sigma-Aldrich

5-Mercuricytidine carbonate 5′-triphosphate triethylammonium salt

~90%

Synonym(s):

Hg-CTP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H15HgN3O14P3 · HCO3
CAS Number:
Molecular Weight:
1148.52
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

~90%

form

solid

storage temp.

−20°C

SMILES string

CCN(CC)CC.NC1=NC(=O)N(C=C1[Hg])[C@@H]2O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O

InChI

1S/C9H15N3O14P3.C6H15N.Hg/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(24-8)3-23-28(19,20)26-29(21,22)25-27(16,17)18;1-4-7(5-2)6-3;/h2,4,6-8,13-14H,3H2,(H,19,20)(H,21,22)(H2,10,11,15)(H2,16,17,18);4-6H2,1-3H3;/t4-,6-,7-,8-;;/m1../s1

InChI key

CKMVNPSLWQKVPG-WFIJOQBCSA-N

Application

5-Mercuricytidine carbonate 5;-triphosphate (Hg-CTP) may be used as a substrate to generate mercurated DNA that may be suitable for image based DNA sequencing strategies.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of deoxyribonucleic acid replication in permeable cells of Bacillus subtilis using mercurated nucleotide substrates.
S Bhattacharya et al.
Biochemistry, 20(11), 3029-3034 (1981-05-26)
Structural analyses on the mercury(II)-mediated T-T base pair.
Tanaka Y, Oda S, et al.
Nucleic Acids Symposium Series, 50, 47-48 (2006)
G Banfalvi et al.
Analytical biochemistry, 146(1), 64-70 (1985-04-01)
A method for isolating picomole quantities of nascent mercurated DNA from a mixture of cellular nucleic acids using affinity chromatography on thiol-agarose is described. Analysis of mercurated DNA (HgDNA) isolated in the presence of in vivo-labeled cellular RNA or in
S Bhattacharya et al.
The Journal of biological chemistry, 257(4), 1610-1612 (1982-02-25)
Mercurated DNA was synthesized in permeable cells of Bacillus subtilis, using 5-mercurideoxycytidine triphosphate as one of the substrates, and was separated from parental unsubstituted DNA by isopycnic centrifugation in CsCl gradients. The ability of mercurated DNA to transform auxotrophic strains
R M Dale et al.
Proceedings of the National Academy of Sciences of the United States of America, 70(8), 2238-2242 (1973-08-01)
A simple acetoxymercuration reaction for introducing covalently bound mercury atoms into nucleotides is described. The 5-mercuriacetate derivatives of UTP, CTP, dUTP, and dCTP, as well as the 7-mercuriacetate derivative of 7-deazaATP, have been prepared by this procedure and tested as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service