Skip to Content
Merck
All Photos(1)

Documents

5.05419

Sigma-Aldrich

α,β-methylene ATP

≥93% (HPLC), powder, P2X purinergic receptor agonist, Calbiochem®

Synonym(s):

α,β-methylene ATP, αβ-MeATP, αβ-methylene ATP, P2X purinergic receptor agonist, α,β-methylene ATP, α,β-Methyleneadenosine 5ʹ-triphosphate lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.77

product name

α,β-methylene ATP,

Assay

≥93% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 100 mg/mL

storage temp.

−20°C

InChI

1S/C11H18N5O12P3.Li/c12-9-6-10(14-2-13-9)16(3-15-6)11-8(18)7(17)5(27-11)1-26-29(19,20)4-30(21,22)28-31(23,24)25;/h2-3,5,7-8,11,17-18H,1,4H2,(H,19,20)(H,21,22)(H2,12,13,14)(H2,23,24,25);/q;+1/p-1/t5-,7-,8-,11-;/m1./s1

InChI key

NVHVREPTGDOYIC-YCSZXMBFSA-M

General description

A metabolically stable and selective P2X purinergic receptor agonist (pEC50 = 5.81) that is shown to be more potent than ATP. Elicits contractions in ileum through the release of endogenous acetylcholine, which can be blocked by tetrodotoxin (~1 mM). Shown to depolarize cells of the longitudinal muscle strips (~ 200 nM to 4 µM) that reaches a peak within 2-3 min after application and then decays to a steady level.

Biochem/physiol Actions

Primary Target
P2X
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Other Notes

Sawyer, G.W., et al. 2000. Br. J. Pharmacol.129, 1458.
Komori, S., et al. 1988. Br. J. Pharmacol.94, 9.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service