EP5409718002
Eppendorf® Rotor Seal and Sealing Grease
for rotors FA-45-20-17 (Centrifuge models 5804/5804R/5810/5810R), and FA-20x5 (Centrifuge models 5920R, 5910R)
Synonym(s):
Eppendorf® Rotor Seal and Sealing Grease
Select a Size
1.590,00 kr.
Select a Size
About This Item
1.590,00 kr.
Recommended Products
packaging
pack of 5 ea
manufacturer/tradename
Eppendorf® 5409718002
compatibility
Centrifuge models 5804/5804R/5810/5810R, and 5920R/5910R
Rotor FA-45-20-17, and FA-20x5
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
5 pcs.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service