Skip to Content
Merck
All Photos(1)

Key Documents

EP5495515003

Eppendorf® Rotor Seal and Sealing Grease

for aerosol-tight rotor lid FA-24×2 (Centrifuge models 5420, 5425/R)

Synonym(s):

Eppendorf® Rotor Lid Seals

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
330,00 kr.

330,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
330,00 kr.

About This Item

UNSPSC Code:
41103913
NACRES:
NB.15

330,00 kr.


Please contact Customer Service for Availability

manufacturer/tradename

Eppendorf® 5495515003

compatibility

Centrifuge models 5420, 5425/R

Looking for similar products? Visit Product Comparison Guide

General description

Seal for aerosol-tight rotor lid FA-24x2/-PTFE (Centrifuge 5425/R)

Application

CENTRIFUGATION

Features and Benefits

FA-24×2 (Centrifuge 5420, 5425/R)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service