Skip to Content
Merck
All Photos(1)

Documents

MABN721

Sigma-Aldrich

Anti-PTK7 Antibody, clone 4F9

ascites fluid, clone 4F9, from mouse

Synonym(s):

Inactive tyrosine-protein kinase 7, CCK-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, Tyrosine-protein kinase-like 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4F9, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTK7(5754)

General description

The gene PTK7 encodes the protein named Inactive tyrosine-protein kinase 7 or Protein-tyrosine kinase 7, or colon carcinoma kinase 4 (CCK4). The PTK7 encoded protein is an interesting single pass type 1 membrane protein involved in Wnt signaling. It is interesting because it contains an inactive tyrosine kinase domain yet it is a component of both the canonical Wnt and non-canonical signaling pathways, and operates during embryogenesis and angiogenesis. Moreover, the PTK7 protein functions in cell migration, cell adhesion, cell polarity, cytoskeleton reorganization and apoptosis, and the PTK7 encoded protein co-localizes with MMP14 at cell junctions as well as the leading edge of migrating cells. The PTK7 encoded protein is expressed in lung, liver, pancreas and other tissues like kidney, placenta and melanocytes. The PTK7 encoded protein does not seem to be expressed in colon and has low expression in brain, heart, and skeletal muscle. EMD-Millipore’s Anti-PTK7 mouse monoclonal, clone 4F9 has been tested by western blot against multiple human cancer cells lines and in immunohistochemistry using paraffin-embedded human lung cancer tissue with DAB detection.

Immunogen

Purified protein fragment of human PTK7 expressed in E. coli.

Application

Detect Protein-tyrosine kinase 7 using this mouse monoclonal antibody, Anti-PTK7 Antibody, clone 4F9 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:100 and 1:500 dilution from a representative lot detected PTK7 in human lung and kidney tissues.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis:A 1:1,000 dilution of this antibody detected PTK7 in 10 µg of A431 cell lysate.

Target description

~130 kDa observed. Uncharacterized bands may appear in some lysate(s). Uniprot describes 6 isoforms between ~90 kDa and ~119 kDa

Physical form

Mouse monoclonal IgG2b ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
A431 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service