B451-100
Tryptic Soy Broth Ready-to-use Bottles
Lecithin 0.5 %, Tween® 4 %, bottle capacity 125 mL, bottle filling volume 100 mL, closure type, white screw cap with septum, box of 12 bottles
Synonym(s):
CASO Broth, Casein Soyabean Digest Broth, TSB, TSB with neutralizer
About This Item
Recommended Products
material
polycarbonate bottle
Quality Level
sterility
sterile; autoclaved
form
powder
feature
closure type white screw cap with septum
composition
Lecithin, 0.5%
Tween®, 4%
packaging
box of 12 bottles
technique(s)
microbiological culture: suitable
sterility testing: suitable
bottle capacity
125 mL
bottle filling volume
100 mL
pH
7.3±0.2 (25 °C)
application(s)
food and beverages
pharmaceutical
storage temp.
2-25°C
suitability
nonselective for bacteria
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Upon incubation, the enumerated microflora can be plated, isolated, and identified.
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service