Skip to Content
Merck
All Photos(2)

Key Documents

1.46610

Millipore

Tryptic Soy Broth Ready-to-use Bottles

bottle capacity 250 mL, bottle filling volume 250 mL, closure type, blue flip cap with septum, box of 6 bottles

Synonym(s):

CASO broth, Soyabean Casein Digest Broth, TSB, TSB with Neutralizer, Tryptic Soy Broth (TSB)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225
Pricing and availability is not currently available.

Agency

EP 2.6.13
JP 4.06
USP 71

Quality Level

sterility

sterile

form

powder

feature

closure type blue flip cap with septum

packaging

box of 6 bottles

bottle capacity

250 mL

bottle filling volume

250 mL

pH

7.3±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for bacteria

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Broth (TSB) is a nutrient rich complex medium designed for the enumeration of total microbial count present in the test sample. The peptones from casein and soymeal to provide enhanced carbon source which promotes growth for the microflora. The broth medium is widely used during sterility testing due to its wide range of specificity.

Application

Tryptic Soy Broth (TSB) is a complex medium used to enumerate microorganisms present in sterile or non-sterile products from food or pharmaceutical industry.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service