Skip to Content
Merck
All Photos(1)

Key Documents

627850

Millipore

Vancomycin, Hydrochloride, Streptomyces orientalis

Amphoteric glycopeptide antibiotic. Inhibits bacterial mucopeptide synthesis by forming complexes.

Synonym(s):

Vancomycin, Hydrochloride, Streptomyces orientalis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H75Cl2N9O24 · xHCl
CAS Number:
Molecular Weight:
1449.25 (free base basis)
UNSPSC Code:
12352200

Quality Level

Assay

≥80% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Amphoteric glycopeptide antibiotic. Inhibits bacterial mucopeptide synthesis by forming complexes. Potency: >700 IU/mg.

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 1 week at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Tsevdou et al.
Foods (Basel, Switzerland), 9(3) (2020-03-25)
The impact of high-pressure (HP) processing on the viability of two probiotic microorganisms (Bifidobacterium bifidum and Lactobacilluscasei) at varying pressure (100-400 MPa), temperature (20-40 °C) and pH (6.5 vs. 4.8) conditions was investigated. Appropriate mathematical models were developed to describe

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service